[5-Amino-2-(4-chlorophenoxy)phenyl]methanol ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Amino-2-(4-chlorophenoxy)benzyl alcohol
047052-500MG
047052-1G
047052-5G
101878-228EA
375.38
USD
101878-228
101878-230
101878-232
[5-Amino-2-(4-chlorophenoxy)phenyl]methanol ≥95%
[5-Amino-2-(4-chlorophenoxy)phenyl]methanol
Hazard: Irritant
Formula:
C₁₃H₁₂ClNO₂ MW: 249.7 g/mol Melting Pt: 129…130 °C |
MDL Number:
MFCD08689719 CAS Number: 924869-26-1 |
Specification Test Results
Melting point | 129-130°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...