N-[4-(tert-Butyl)phenyl]-N'-(2-chloroethyl)urea ≥95%

Supplier: Matrix Scientific

047050-500MG 047050-1G 047050-5G
101878-216EA 278.46 USD
101878-216 101878-218 101878-220
N-[4-(tert-Butyl)phenyl]-N'-(2-chloroethyl)urea ≥95%
N-[4-(tert-Butyl)phenyl]-N'-(2-chloroethyl)urea

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O
MW: 254.76 g/mol
Melting Pt: 126…128 °C
MDL Number: MFCD00933173
CAS Number: 118202-59-8

Order Now

Specification Test Results

Melting point 126-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR