2-(4-Hydroxy-2-phenyl-1,3-thiazol-5-yl)acetic acid ≥95%

Supplier: Matrix Scientific

047049-500MG 047049-1G 047049-5G
101878-210EA 181.54 USD
101878-210 101878-212 101878-214
2-(4-Hydroxy-2-phenyl-1,3-thiazol-5-yl)acetic acid ≥95%
2-(4-Hydroxy-2-phenyl-1,3-thiazol-5-yl)acetic acid

Hazard: Irritant

Formula: C₁₁H₉NO₃S
MW: 235.26 g/mol
Melting Pt: 196…200 °C (dec.)
MDL Number: MFCD08689717
CAS Number: 133834-03-4

Order Now

Specification Test Results

Melting point 196-200°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR