N-(2-Chloroethyl)-N'-(4-methoxyphenyl)urea ≥95%

Supplier: Matrix Scientific

047048-500MG 047048-1G 047048-5G
101878-204EA 181.54 USD
101878-204 101878-206 101878-208
N-(2-Chloroethyl)-N'-(4-methoxyphenyl)urea ≥95%
N-(2-Chloroethyl)-N'-(4-methoxyphenyl)urea

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O₂
Melting Pt: 156…158 °C
MDL Number: MFCD02671069
CAS Number: 65536-40-5

Order Now

Specification Test Results

Melting point 156-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR