N-(2-Chloroethyl)-N'-[4-(trifluoromethoxy)phenyl]urea ≥95%

Supplier: Matrix Scientific

047047-500MG 047047-1G 047047-5G
101878-198EA 278.46 USD
101878-198 101878-200 101878-202
N-(2-Chloroethyl)-N'-[4-(trifluoromethoxy)phenyl]urea ≥95%
N-(2-Chloroethyl)-N'-[4-(trifluoromethoxy)phenyl]urea

Hazard: Irritant

Formula: C₁₀H₁₀ClF₃N₂O₂
MW: 282.65 g/mol
Melting Pt: 121…123 °C
MDL Number: MFCD08689716
CAS Number: 937601-75-7

Order Now

Specification Test Results

Melting point 121-123°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR