3-Chloro-6-nitro-1H-indazole ≥95%

Supplier: Matrix Scientific

047046-500MG 047046-1G 047046-5G
101878-192EA 278.46 USD
101878-192 101878-194 101878-196
3-Chloro-6-nitro-1H-indazole ≥95%
3-Chloro-6-nitro-1H-indazole

Hazard: Irritant

Formula: C₇H₄ClN₃O₂
MW: 197.5800 g/mol
Melting Pt: 203-205°C
MDL Number: MFCD00010741
CAS Number: 50593-68-5

Order Now

Specification Test Results

Melting point 203-205°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR