2-(3-Nitrophenyl)nicotinic acid ≥95%

Supplier: Matrix Scientific

047035-500MG 047035-1G 047035-5G
101878-150EA 375.38 USD
101878-150 101878-152 101878-154
2-(3-Nitrophenyl)nicotinic acid ≥95%
2-(3-Nitrophenyl)nicotinic acid

Hazard: Irritant

Formula: C₁₂H₈N₂O₄
Melting Pt: 214-216°C
MDL Number: MFCD08689711
CAS Number: 937601-72-4

Order Now

Specification Test Results

Melting point 214-216°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR