Methyl-4'-methoxy-4-nitro[1,1'-biphenyl]-2-carboxylate ≥95%

Supplier: Matrix Scientific

047033-500MG 047033-1G 047033-5G
101878-138EA 278.46 USD
101878-138 101878-140 101878-142
Methyl-4'-methoxy-4-nitro[1,1'-biphenyl]-2-carboxylate ≥95%
Methyl-4'-methoxy-4-nitro[1,1'-biphenyl]-2-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₃NO₅
MW: 287.27 g/mol
Melting Pt: 69…71 °C
MDL Number: MFCD08689709
CAS Number: 937601-71-3

Order Now

Specification Test Results

Melting point 69-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR