2-Amino-6,8-dibromo-3-quinolinecarboxamide ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Amino-6,8-dibromoquinoline-3-carboxamide
047032-500MG
047032-1G
047032-5G
101878-132EA
130.77
USD
101878-132
101878-134
101878-136
2-Amino-6,8-dibromo-3-quinolinecarboxamide ≥95%
2-Amino-6,8-dibromo-3-quinolinecarboxamide
Hazard: Irritant
Formula:
C₁₀H₇Br₂N₃O MW: 345 g/mol Melting Pt: 283…286 °C |
MDL Number:
MFCD08689708 CAS Number: 937601-68-8 |
Specification Test Results
Melting point | 283-286°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...