Methyl-4-amino-3-fluorobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-amino-3-fluorobenzoate

047030-1G 047030-5G 047030-25G
101878-120EA 15.38 USD
101878-120 101878-122 101878-124
Methyl-4-amino-3-fluorobenzoate ≥95%
Methyl 4-amino-3-fluorobenzoate

Hazard: Irritant

Formula: C₈H₈FNO₂
MW: 169.1530 g/mol
Melting Pt: 107…110 °C
MDL Number: MFCD08689703
CAS Number: 185629-32-7

Order Now

Specification Test Results

Melting point 107-110°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR