6,8-Dichloro-3-(4-nitrophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%

Supplier: Matrix Scientific

047028-500MG 047028-1G 047028-5G
101878-108EA 181.54 USD
101878-108 101878-110 101878-112
6,8-Dichloro-3-(4-nitrophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
6,8-Dichloro-3-(4-nitrophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone

Hazard: Irritant

Formula: C₁₄H₇Cl₂N₃O₃S
MW: 368.2 g/mol
Melting Pt: 282…284 °C
MDL Number: MFCD08689701
CAS Number: 937601-65-5

Order Now

Specification Test Results

Melting point 282-284°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR