3-(4-Bromophenyl)-5-methyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
Supplier: Matrix Scientific
047027-500MG
047027-1G
047027-5G
101878-102EA
181.54
USD
101878-102
101878-104
101878-106
3-(4-Bromophenyl)-5-methyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
3-(4-Bromophenyl)-5-methyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Hazard: Irritant
Formula:
C₁₅H₁₁BrN₂OS MW: 347.24 g/mol Melting Pt: 344…346 °C |
MDL Number:
MFCD08689700 CAS Number: 937601-63-3 |
Specification Test Results
Melting point | 344-346°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...