3-Fluoro-4-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

047023-500MG 047023-1G 047023-5G
101878-078EA 40 USD
101878-078 101878-080 101878-082
3-Fluoro-4-nitrobenzaldehyde ≥95%
3-Fluoro-4-nitrobenzaldehyde

Hazard: Irritant

Formula: C₇H₄NO₃F
MW: 169.1100 g/mol
Melting Pt: 57-58°C
MDL Number: MFCD00968940
CAS Number: 160538-51-2

Order Now

Specification Test Results

Melting point 57-58°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR