3-(4-Nitrophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%

Supplier: Matrix Scientific

047021-500MG 047021-1G 047021-5G
101878-066EA 181.54 USD
101878-066 101878-068 101878-070
3-(4-Nitrophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
3-(4-Nitrophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone

Hazard: Irritant

Formula: C₁₄H₉N₃O₃S
MW: 299.31 g/mol
Melting Pt: 319…320 °C
MDL Number: MFCD02916354
CAS Number: 72176-80-8

Order Now

Specification Test Results

Melting point 319-320°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR