3-(2-Bromo-4-fluorophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
Supplier: Matrix Scientific
047017-500MG
047017-1G
047017-5G
101878-044EA
181.54
USD
101878-044
101878-046
101878-048
3-(2-Bromo-4-fluorophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
3-(2-Bromo-4-fluorophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Hazard: Irritant
Formula:
C₁₄H₈BrFN₂OS Melting Pt: 267…269 °C |
MDL Number:
MFCD08689698 CAS Number: 937601-60-0 |
Specification Test Results
Melting point | 267-269°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...