3-Fluoro-4-nitrobenzylalcohol ≥95%

Supplier: Matrix Scientific

Synonyms: (3-Fluoro-4-nitrophenyl)methanol

047014-500MG 047014-1G 047014-5G
101878-026EA 89.23 USD
101878-026 101878-028 101878-030
3-Fluoro-4-nitrobenzylalcohol ≥95%
3-Fluoro-4-nitrobenzylalkohol

Hazard: Irritant

Formula: C₇H₆FNO₃
MW: 171.1300 g/mol
Melting Pt: 96…97 °C
MDL Number: MFCD03094236
CAS Number: 503315-74-0

Order Now

Specification Test Results

Melting point 96-97°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR