2-Cyano-N-[4-(trifluoromethyl)benzyl]acetamide ≥95%

Supplier: Matrix Scientific

047013-500MG 047013-1G 047013-5G
101878-020EA 181.54 USD
101878-020 101878-022 101878-024
2-Cyano-N-[4-(trifluoromethyl)benzyl]acetamide ≥95%
2-Cyano-N-[4-(trifluoromethyl)benzyl]acetamide

Hazard: Irritant

Formula: C₁₁H₉F₃N₂O
Melting Pt: 125…127 °C
MDL Number: MFCD08689696
CAS Number: 566926-08-7

Order Now

Specification Test Results

Melting point 125-127°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR