6-Chloro-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Chloro-2,3-dihydro-3-phenyl-2-thioxo-1H-quinazolin-4-one
047004-500MG
047004-1G
047004-5G
101877-986EA
181.54
USD
101877-986
101877-988
101877-990
6-Chloro-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
6-Chloro-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Hazard: Irritant
Formula:
C₁₄H₉ClN₂OS MW: 288.76 g/mol Melting Pt: >350 °C |
MDL Number:
MFCD02650967 CAS Number: 84772-27-0 |
Specification Test Results
Melting point | >350°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...