7-Chloro-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
Supplier: Matrix Scientific
047003-500MG
047003-1G
047003-5G
101877-980EA
181.54
USD
101877-980
101877-982
101877-984
7-Chloro-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
7-Chloro-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Hazard: Irritant
Formula:
C₁₄H₉ClN₂OS MW: 288.76 g/mol Melting Pt: 323…324 °C |
MDL Number:
MFCD02650968 CAS Number: 13165-15-6 |
Specification Test Results
Melting point | 323-324°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...