1-Benzyl-2-(4-isopropylphenyl)-6-oxo-3-piperidinecarboxylic acid ≥95%
Supplier: Matrix Scientific
047000-500MG
047000-1G
047000-5G
101877-974EA
278.46
USD
101877-974
101877-976
101877-978
1-Benzyl-2-(4-isopropylphenyl)-6-oxo-3-piperidinecarboxylic acid ≥95%
1-Benzyl-2-(4-isopropylphenyl)-6-oxo-3-piperidinecarboxylic acid
Hazard: Irritant
Formula:
C₂₂H₂₅NO₃ Melting Pt: 164…165 °C |
MDL Number:
MFCD09027094 CAS Number: 937604-04-1 |
Specification Test Results
Melting point | 164-165°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...