6-Bromoquinazoline-2,4(1H,3H)-dione ≥95%
Supplier: Matrix Scientific
046995-1G
046995-5G
046995-25G
101877-950EA
44.62
USD
101877-950
101877-952
101877-954
6-Bromoquinazoline-2,4(1H,3H)-dione ≥95%
6-Bromoquinazoline-2,4(1H,3H)-dione
Hazard: Irritant
Formula:
C₈H₅BrN₂O₂ MW: 241.04 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00462868 CAS Number: 88145-89-5 |
Specification Test Results
Melting point | >300°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...