1-Benzyl-2-(4-methoxyphenyl)-6-oxo-3-piperidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

046993-500MG 046993-1G 046993-5G
101877-944EA 278.46 USD
101877-944 101877-946 101877-948
1-Benzyl-2-(4-methoxyphenyl)-6-oxo-3-piperidinecarboxylic acid ≥95%
1-Benzyl-2-(4-methoxyphenyl)-6-oxo-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₂₀H₂₁NO₄
Melting Pt: 146…148 °C
MDL Number: MFCD07377836
CAS Number: 937604-06-3

Order Now

Specification Test Results

Melting point 146-148°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR