2-Acetyl-6-bromopyridine ≥95%

Supplier: Matrix Scientific

046992-1G 046992-5G
101877-940EA 18.46 USD
101877-940 101877-942
2-Acetyl-6-bromopyridine ≥95%
2-Acetyl-6-bromopyridine

Hazard: Irritant

Formula: C₇H₆BrNO
MW: 200.03 g/mol
Melting Pt: 55…57 °C
MDL Number: MFCD00272200
CAS Number: 49669-13-8

Order Now

Specification Test Results

Melting point 55-57°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR