Methyl (2S)-1-(5-nitro-2-pyridinyl)tetrahydro-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl (2S)-1-(5-nitropyridin-2-yl)pyrrolidine-2-carboxylate

046990-500MG 046990-1G 046990-5G
101877-926EA 278.46 USD
101877-926 101877-928 101877-930
Methyl (2S)-1-(5-nitro-2-pyridinyl)tetrahydro-1H-pyrrole-2-carboxylate ≥95%
Methyl (2S)-1-(5-nitro-2-pyridinyl)tetrahydro-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₄
MW: 251.24 g/mol
Melting Pt: 87…88 °C
MDL Number: MFCD08689691
CAS Number: 122092-22-2

Order Now

Specification Test Results

Melting point 87-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR