3-Iodo-4-nitro-1H-indazole ≥95%

Supplier: Matrix Scientific

046986-500MG 046986-1G
101877-910EA 556.92 USD
101877-910 101877-912
3-Iodo-4-nitro-1H-indazole ≥95%
3-Iodo-4-nitro-1H-indazole

Hazard: Irritant

Formula: C₇H₄IN₃O₂
Melting Pt: 210-212°C
MDL Number: MFCD07781574
CAS Number: 885521-22-2

Order Now

Specification Test Results

Melting point 210-212°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR