3-Bromo-1-methyl-6-nitro-1H-indazole ≥95%
Supplier: Matrix Scientific
046982-500MG
046982-1G
101877-898EA
921.54
USD
101877-898
101877-900
3-Bromo-1-methyl-6-nitro-1H-indazole ≥95%
3-Bromo-1-methyl-6-nitro-1H-indazole
Hazard: Irritant
Formula:
C₈H₆BrN₃O₂ Melting Pt: 154-156°C |
MDL Number:
MFCD12025932 CAS Number: 74209-32-8 |
Specification Test Results
Melting point | 154-156°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...