3-(1H-Pyrrol-1-yl)-1-benzothiophene-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

046978-500MG 046978-1G
101877-886EA 556.92 USD
101877-886 101877-888
3-(1H-Pyrrol-1-yl)-1-benzothiophene-2-carboxylic acid ≥95%
3-(1H-Pyrrol-1-yl)-1-benzothiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₉NO₂S
MW: 243.29 g/mol
Melting Pt: 207…209 °C
MDL Number: MFCD12025930
CAS Number: 80066-98-4

Order Now

Specification Test Results

Melting point 207-209°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR