5-Chloro-1-benzothiophene-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
046976-500MG
046976-1G
101877-878EA
480
USD
101877-878
101877-880
5-Chloro-1-benzothiophene-2-carboxylic acid ≥95%
5-Chloro-1-benzothiophene-2-carboxylic acid
Hazard: Irritant
Formula:
C₉H₅ClO₂S Melting Pt: 256…258 °C |
MDL Number:
MFCD01927183 CAS Number: 13771-75-0 |
Specification Test Results
Melting point | 256-258°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...