2,3,5-Trichloro-4-iodopyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Iodo-2,3,5-trichloropyridine
046974-500MG
046974-1G
046974-5G
101877-866EA
343.08
USD
101877-866
101877-868
101877-870
2,3,5-Trichloro-4-iodopyridine ≥95%
2,3,5-Trichloro-4-iodopyridine
Hazard: Irritant
Formula:
C₅HCI₃IN Melting Pt: 121-122°C |
CAS Number:
406676-23-1 |
Specification Test Results
Melting point | 121-122°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...