Methyl-4-chloro-1H-pyrrolo[3,2-c]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-Chloro-5-azaindole-2-carboxylate

046973-500MG 046973-1G
101877-862EA 921.54 USD
101877-862 101877-864
Methyl-4-chloro-1H-pyrrolo[3,2-c]pyridine-2-carboxylate ≥95%
Methyl-4-chloro-1H-pyrrolo[3,2-c]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₉H₇ClN₂O₂
MW: 210.62 g/mol
Melting Pt: 224…225 °C
MDL Number: MFCD09870065
CAS Number: 688357-19-9

Order Now

Specification Test Results

Melting point 224-225°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR