6-Bromo-2-oxo-1,2-dihydro-3-quinolinecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-1,2-dihydro-2-oxoquinoline-3-carbonitrile

046972-500MG 046972-1G
101877-858EA 412.31 USD
101877-858 101877-860
6-Bromo-2-oxo-1,2-dihydro-3-quinolinecarbonitrile ≥95%
6-Bromo-2-oxo-1,2-dihydro-3-quinolinecarbonitrile

Hazard: Irritant

Formula: C₁₀H₅BrN₂O
MW: 249.07 g/mol
Melting Pt: >300 °C
MDL Number: MFCD09724162
CAS Number: 99465-03-9

Order Now

Specification Test Results

Melting point >300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR