2,6-Dichloro-4-(trifluoromethyl)nicotinic acid ≥95%
Supplier: Matrix Scientific
046971-1G
046971-5G
101877-854EA
861.54
USD
101877-854
101877-856
2,6-Dichloro-4-(trifluoromethyl)nicotinic acid ≥95%
2,6-Dichloro-4-(trifluoromethyl)nicotinic acid
Hazard: Irritant
Formula:
C₇H₂Cl₂F₃NO₂ Melting Pt: 79…80 °C |
MDL Number:
MFCD06208180 CAS Number: 503437-19-2 |
Specification Test Results
Melting point | 79-80°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...