2-Amino-5-bromobenzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromoanthranilaldehyde

046968-500MG 046968-1G
101877-842EA 69.23 USD
101877-842 101877-844
2-Amino-5-bromobenzaldehyde ≥95%
2-Amino-5-bromobenzaldehyde

Hazard: Irritant

Formula: C₇H₆BrNO
MW: 200.00 g/mol
Boiling Pt: 290 °C
Melting Pt: 75…77 °C
Density: 1.673
Flash Pt: 129 °C
MDL Number: MFCD10696879
CAS Number: 29124-57-0

Order Now

Specification Test Results

Melting point 75-77°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR