N-(5-Amino-2-hydroxyphenyl)acetamide ≥95%
Supplier: Matrix Scientific
046967-500MG
046967-1G
101877-838EA
375.38
USD
101877-838
101877-840
N-(5-Amino-2-hydroxyphenyl)acetamide ≥95%
N-(5-Amino-2-hydroxyphenyl)acetamide
Hazard: Irritant
Formula:
C₈H₁₀N₂O₂ Melting Pt: 162 °C |
MDL Number:
MFCD12025926 CAS Number: 23184-60-3 |
Specification Test Results
Melting point | 162°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...