2-Chloro-N-(2-hydroxy-5-nitrophenyl)acetamide ≥95%

Supplier: Matrix Scientific

046966-500MG 046966-1G 046966-5G
101877-832EA 181.54 USD
101877-832 101877-834 101877-836
2-Chloro-N-(2-hydroxy-5-nitrophenyl)acetamide ≥95%
2-Chloro-N-(2-hydroxy-5-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₈H₇ClN₂O₄
Melting Pt: 245 °C
MDL Number: MFCD01006663
CAS Number: 35588-39-7

Order Now

Specification Test Results

Melting point 245°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR