Methyl-5-chloro-6-fluoronicotinate ≥95%

Supplier: Matrix Scientific

046965-500MG 046965-1G
101877-828EA 308.58 USD
101877-828 101877-830
Methyl-5-chloro-6-fluoronicotinate ≥95%
Methyl-5-chloro-6-fluoronicotinate

Hazard: Irritant

Formula: C₇H₅ClFNO₂
Melting Pt: 71…72 °C
MDL Number: MFCD12498696
CAS Number: 1171918-85-6

Order Now

Specification Test Results

Melting point 71-72°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR