(5-Chloro-2-pyridinyl)methanol ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Chloro-2-(hydroxymethyl)pyridine, (5-Chloropyridin-2-yl)methanol
046962-500MG
046962-1G
101877-816EA
69.23
USD
101877-816
101877-818
(5-Chloro-2-pyridinyl)methanol ≥95%
(5-Chloro-2-pyridinyl)methanol
Hazard: Irritant
Formula:
C₆H₆ClNO MW: 143.5709 g/mol Melting Pt: 45…47 °C |
MDL Number:
MFCD10000856 CAS Number: 209526-98-7 |
Specification Test Results
Melting point | 45-47°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...