5,6-Dihydro-4H-cyclopenta[b]thiophene-2-carbohydrazide ≥95%
Supplier: Matrix Scientific
046959-500MG
046959-1G
101877-806EA
278.46
USD
101877-806
101877-808
5,6-Dihydro-4H-cyclopenta[b]thiophene-2-carbohydrazide ≥95%
5,6-Dihydro-4H-cyclopenta[b]thiophene-2-carbohydrazide
Hazard: Irritant
Formula:
C₈H₁₀N₂OS Melting Pt: 154…156 °C |
MDL Number:
MFCD06335037 CAS Number: 750599-01-0 |
Specification Test Results
Melting point | 154-156°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...