Ethyl-2-[(tert-butoxycarbonyl)amino]-4-(trifluoromethyl)-1,3-oxazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-amino-4-(trifluoromethyl)-1,3-oxazole-5-carboxylate, 2-BOC protected

046958-500MG 046958-1G
101877-802EA 375.38 USD
101877-802 101877-804
Ethyl-2-[(tert-butoxycarbonyl)amino]-4-(trifluoromethyl)-1,3-oxazole-5-carboxylate ≥95%
Ethyl-2-[(tert-butoxycarbonyl)amino]-4-(trifluoromethyl)-1,3-oxazole-5-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₅F₃N₂O₅
MW: 324.26 g/mol
Melting Pt: 76…78 °C
MDL Number: MFCD12827772
CAS Number: 1186405-12-8

Order Now

Specification Test Results

Melting point 76-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR