4-Fluoro-1-benzothiophene-2-carbohydrazide ≥95%

Supplier: Matrix Scientific

046955-500MG 046955-1G 046955-5G
101877-788EA 375.38 USD
101877-788 101877-790 101877-792
4-Fluoro-1-benzothiophene-2-carbohydrazide ≥95%
4-Fluoro-1-benzothiophene-2-carbohydrazide

Hazard: Irritant

Formula: C₉H₇FN₂OS
MW: 210.23 g/mol
Melting Pt: 196…198 °C
MDL Number: MFCD11643395
CAS Number: 1098351-44-0

Order Now

Specification Test Results

Melting point 196-198°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR