2-(Trifluoromethyl)imidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

046950-500MG 046950-1G
101877-766EA 300 USD
101877-766 101877-768
2-(Trifluoromethyl)imidazo[1,2-a]pyridine ≥95%
2-(Trifluoromethyl)imidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₅F₃N₂
MDL Number: MFCD11111687
CAS Number: 73221-12-2

Order Now

Specification Test Results

Melting point 90-92°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR