6-Bromo-4-oxo-1,4-dihydro-8-quinolinecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-1,4-dihydro-4-oxoquinoline-8-carbonitrile

046943-500MG 046943-1G
101877-734EA 369.23 USD
101877-734 101877-736
6-Bromo-4-oxo-1,4-dihydro-8-quinolinecarbonitrile ≥95%
6-Bromo-4-oxo-1,4-dihydro-8-quinolinecarbonitrile

Hazard: Irritant

Formula: C₁₀H₅BrN₂O
MW: 249.07 g/mol
Melting Pt: >300 °C
MDL Number: MFCD12025922
CAS Number: 1160474-80-5

Order Now

Specification Test Results

Melting point >300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR