5-Methoxy-4-piperazino-2-(trifluoromethyl)pyrimidine ≥95%
Supplier: Matrix Scientific
046940-500MG
046940-1G
101877-722EA
921.54
USD
101877-722
101877-724
5-Methoxy-4-piperazino-2-(trifluoromethyl)pyrimidine ≥95%
5-Methoxy-4-piperazino-2-(trifluoromethyl)pyrimidine
Hazard: Irritant
Formula:
C₁₀H₁₃F₃N₄O MW: 262.24 g/mol Melting Pt: 101…102 °C |
MDL Number:
MFCD11100216 CAS Number: 1048912-73-7 |
Specification Test Results
Melting point | 101-102°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...