N-(2-Hydroxyethyl)-2-nitrobenzenesulfonamide ≥95%
Supplier: Matrix Scientific
046939-500MG
046939-1G
046939-5G
101877-716EA
170.77
USD
101877-716
101877-718
101877-720
N-(2-Hydroxyethyl)-2-nitrobenzenesulfonamide ≥95%
N-(2-Hydroxyethyl)-2-nitrobenzenesulfonamide
Hazard: Irritant
Formula:
C₈H₁₀N₂O₅S Melting Pt: 83…85 °C |
MDL Number:
MFCD00458943 CAS Number: 18226-11-4 |
Specification Test Results
Melting point | 83-85°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...