2-(1,3-Benzothiazol-2-ylamino)-1-ethanol ≥95%

Supplier: Matrix Scientific

046938-500MG 046938-1G 046938-5G
101877-710EA 278.46 USD
101877-710 101877-712 101877-714
2-(1,3-Benzothiazol-2-ylamino)-1-ethanol ≥95%
2-(1,3-Benzothiazol-2-ylamino)-1-ethanol

Hazard: Irritant

Formula: C₉H₁₀N₂OS
Melting Pt: 102…104 °C
MDL Number: MFCD00519113
CAS Number: 18392-47-7

Order Now

Specification Test Results

Melting point 102-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR