6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carbohydrazide ≥95%

Supplier: Matrix Scientific

046936-500MG 046936-1G
101877-700EA 556.92 USD
101877-700 101877-702
6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carbohydrazide ≥95%
6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carbohydrazide

Hazard: Irritant

Formula: C₉H₇F₃N₄O
Melting Pt: 202…203 °C
MDL Number: MFCD11100215
CAS Number: 1048912-65-7

Order Now

Specification Test Results

Melting point 202-203°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR