5-Bromo-6-cyclopropyl-2-hydroxy-4-(trifluoromethyl)nicotinonitrile ≥95%
Supplier: Matrix Scientific
046935-500MG
046935-1G
101877-696EA
747.69
USD
101877-696
101877-698
5-Bromo-6-cyclopropyl-2-hydroxy-4-(trifluoromethyl)nicotinonitrile ≥95%
5-Bromo-6-cyclopropyl-2-hydroxy-4-(trifluoromethyl)nicotinonitrile
Hazard: Irritant
Formula:
C₁₀H₆BrF₃N₂O Melting Pt: 219…220 °C |
MDL Number:
MFCD11553027 CAS Number: 1092352-29-8 |
Specification Test Results
Melting point | 219-220°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...