4-Chloro-N-(2-hydroxyethyl)benzenesulfonamide ≥95%
Supplier: Matrix Scientific
046933-500MG
046933-1G
046933-5G
101877-686EA
181.54
USD
101877-686
101877-688
101877-690
4-Chloro-N-(2-hydroxyethyl)benzenesulfonamide ≥95%
4-Chloro-N-(2-hydroxyethyl)benzenesulfonamide
Hazard: Irritant
Formula:
C₈H₁₀ClNO₃S Melting Pt: 106…108 °C |
MDL Number:
MFCD01185491 CAS Number: 6419-69-8 |
Specification Test Results
Melting point | 106-108°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...