4-Nitro-1-isoindolinone ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Nitroisoindolin-1-one
046930-500MG
046930-1G
101877-678EA
556.92
USD
101877-678
101877-680
4-Nitro-1-isoindolinone ≥95%
4-Nitro-1-isoindolinone
Hazard: Irritant
Formula:
C₈H₆N₂O₃ MW: 178.15 g/mol Melting Pt: 225…230 °C |
MDL Number:
MFCD09701290 CAS Number: 366452-97-3 |
Specification Test Results
Melting point | 225-230°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...