5-Methoxy-2-(trifluoromethyl)-4-pyrimidinol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-5-methoxy-2-(trifluoromethyl)pyrimidine

046929-500MG 046929-1G
101877-674EA 556.92 USD
101877-674 101877-676
5-Methoxy-2-(trifluoromethyl)-4-pyrimidinol ≥95%
5-Methoxy-2-(trifluoromethyl)-4-pyrimidinol

Hazard: Irritant

Formula: C₆H₅F₃N₂O₂
MW: 194.11 g/mol
Melting Pt: 225…227 °C
MDL Number: MFCD11100214
CAS Number: 808770-42-5

Order Now

Specification Test Results

Melting point 225-227°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR